Key Documents
Z756857
DNA band excisor
pkg of 10 ea
Synonym(s):
electrophoresis gels, gel extraction
About This Item
UNSPSC Code:
41106300
material
polystyrene
packaging
pkg of 10 ea
manufacturer/tradename
Bel-Art F36970-0010
General description
Safely excise DNA bands while protecting your hands and transilluminator. Double-ended polystyrene tool with thin blade and spatula is perfect for excising bands from agarose or polyacrylamide gels with precision and placing into test tubes.
  • Excise DNA bands as well as purified plasmid DNA preps from electrophoresis gels
  • Polystyrene tool will not cut fingers or scratch transilluminator screens
  • Use tool to speed up processing time, reducing exposure of nucleic acids and user′s hands to damaging UV light
  • Discard in standard trash receptacles; no sharps container required
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1