Key Documents
Z756849
In-Line equilibrium cell
pack of 1 ea, clear acrylic
About This Item
UNSPSC Code:
41123100
NACRES:
NB.22
material
clear acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H40317-0000
General description
The in-line equilibrium cell has a top opening for addition or removal of solutions and can contain 1 mL in every half cell. PTFE-coated stainless steel screws secure the cell.
  • Cell dimensions: 152 mm L x 25 mm W x 76 mm H (6 in. x 1 in. x 3 in.)
  • 5 in-line cavities
  • Membrane is secured by 7 stainless steel screws with wing nuts
Choose from one of the most recent versions:
Certificates of Analysis (COA)