Key Documents
Z756822
Equilibrium type dialysis cell
capacity 5 mL, pack of 1 ea
Synonym(s):
dialysis tubes
About This Item
UNSPSC Code:
41123100
NACRES:
NB.22
material
clear acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H40261-0000
capacity
5 mL
pore size
≥6 kDa MWCO
General description
These dialysis units feature stainless steel screws which secure the membrane acting as a seal. PTFE-coated stainless steel screws are located in the filler holes. Volume displayed is for a single half-cell. Total cell capacity is 2x of that shown. A red dot is featured on one half of the dialysis cell for easily identifying otherwise identical halves.
Choose from one of the most recent versions:
Certificates of Analysis (COA)