Key Documents
Z756474
PrepSafe replacement vortexing attachment
pkg of 1 ea
Synonym(s):
rack test tube, test tube racks, tube racks
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F18742-0140
General description
Replacement vortexing attachment snaps into PrepSafe microcentrifuge tube mini floating racks, which permits vortex mixing of tubes.
Legal Information
PrepSafe is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)