Z756296
BRAND® BIO-CERT® filter tips, Tip-Box N
low retention, volume range 5-100 μL
Synonym(s):
pipette filter tips, pipette tips, pipettor tips
About This Item
material
colorless
sterility
sterile
feature
ATP free
DNA free
RNase free
barrier
endotoxin free
filter
low retention
packaging
pack of 960 ea
manufacturer/tradename
BRAND 703159
maximum volume
100 μL
volume range
5-100 μL
Looking for similar products? Visit Product Comparison Guide
General description
- High-purity polypropylene type
- Manufactured without lubricants
- Cadmium-free pigments
- Autoclavable at 121°C/250°F (2 bar) acc. DIN EN 285
- Environmentally friendly packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service