Key Documents
Z744072
EZcap General Purpose FilmStrips
polypropylene, non-sterile
Select a Size
400 EACH
$34.40
$34.40
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
400 EACH
$34.40
About This Item
UNSPSC Code:
41122410
NACRES:
NB.04
$34.40
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
material
polypropylene
sterility
non-sterile
feature
RNase and DNase free
packaging
case of 400 ea
manufacturer/tradename
Excel Scientific GP-CS1X8-400
parameter
-40 °C temp. range (120 °C)
Legal Information
EZcap is a trademark of Excel Scientific, Inc.
FilmStrips is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)