Key Documents
Z743530
Petri-Sleeve®
About This Item
UNSPSC Code:
55121611
manufacturer/tradename
Diversified Biotech PTSV-1000
roll L × W
7 in. × 50 ft , 3 in white square printed every 6 in
General description
7" wide, 2mil thick polytubing on a 50′ continuous roll with 3" white squares printed every 6".
Ideal Uses: Sealing pre-loaded Petri dishes for short and medium term storage.
ideal for sealing pre-loaded Petri dishes for short and medium term storage. Petri-Sleeve can be sealed using standard laboratory tape or an impulse sealer.
Legal Information
Petri-Sleeve is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)