Key Documents
Z743519
Perf-Tape perforated labeling tape
blue, L × W 0.75 in. × 2160 in. , with 1.25 in. perforations
About This Item
UNSPSC Code:
55121611
manufacturer/tradename
Diversified Biotech LBTP-4300
L × W
0.75 in. × 2160 in. , with 1.25 in. perforations
color
blue
General description
0.75" durable paper laboratory labeling tape with rubber adhesive on a 1" plastic core with perforations every 1.25". Perforations can be left together to create larger pieces when needed. For example, tape can be sized at 1.25", 2.50", 3.75" etc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)