Z743465
DuraSeal™ cling laboratory stretch film
size 2 in. × 500 ft
Synonym(s):
heat-resistant stretch film, laboratory cling film, plastic stretch film, solvent-resistant stretch film
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Recommended Products
manufacturer/tradename
Diversified Biotech DSC2-500
size
2 in. × 500 ft
Looking for similar products? Visit Product Comparison Guide
General description
It Clings- Film can cling to itself and glass vessels
Heat Resistant - Softens at 100°C
Strong- Film can stretch 300-400% before breaking or tearing
Tight Sealing?- Forms an air tight barrier when stretched across openings
Versatile - Compatibile in more applications than Parafilm®
Heat Resistant - Softens at 100°C
Strong- Film can stretch 300-400% before breaking or tearing
Tight Sealing?- Forms an air tight barrier when stretched across openings
Versatile - Compatibile in more applications than Parafilm®
Legal Information
DuraSeal is a trademark of Diversified Biotech
Parafilm is a registered trademark of Bemis Company, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service