Key Documents
Z742243
IBI Blot Washer
AC/DC input 115 V AC, US 3-pin plug
About This Item
UNSPSC Code:
12352200
size
9.0 in. × 9.75 in. × 5.5 in.
weight
8 lb
General description
The IBI Blot Washer system provides an economical way to automate your western blotting procedures while reducing background issues. The pump/control unit coupled with the 4 L wash and waste reservoirs provide automated dispensing and aspiration of western blot reagents.
The blot washer pump/control unit contains a microprocessor and software that allows for the development of up to 20 stored programs. The software allows you to control all elements of the instruments capability: volume of wash or other reagent dispensed, number and duration of washes, and aspiration of reagents to waste. The user-friendly software offers an intuitive approach and allows the user to program a procedure in minutes.
The dispensing and aspirate manifolds are made up of hard plastic tubes that can be fastened to the side of the blot box and easily adjusted for any depth to accommodate any existing blot boxes in the lab.
Accessories include a quad harness which allows for processing up to four blots at once and a 140 ml syringe which can be added for secondary reagent dispensing.
Features and Benefits
  • Automated dispensing and aspiration of blot wash reagents
  • Programmable washing time, liquid dispense rate/volume, and aspiration rate/volume
  • Comes with: the pump/module, 4L wash bottle with cap, 4L waste bottle with cap, mini blot tray, standard blot tray, vacuum tube with connectors, dispense manifold, aspirate manifold, power cord, and operation manual
Choose from one of the most recent versions:
Certificates of Analysis (COA)