Key Documents
Z742103
Cell Strainer
white, Individually Wrapped, sterile; γ-irradiated, pack of 50 ea
About This Item
UNSPSC Code:
41104932
NACRES:
NB.22
material
Nylon
white
sterility
sterile; γ-irradiated
packaging
pack of 50 ea
pkg of (individually wrapped)
manufacturer/tradename
MIDSCI SG-70ICS
pore size
70 μm
General description
Cell Strainers from MIDSCI can be used for many applications including isolating cells and removing clumps or debris. Cell Strainers from MIDSCI are compatible with standard 50mL centrifuge tubes. The exclusive Pestle design improves manipulation of material in the strainer during operation.
  • Polypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materials
  • Elongated lip on housing extends beyond edge of centrifuge tube enabling aseptic handling with forceps
  • Extended gripping tab on housing for easy handling
  • Available in three porosities: 40, 70, 100 um
  • Color coded by mesh porosity for easy identification
  • Packaged sterile (gamma irradiated) in individually wrapped peel-to-open paper/plastic wrapper or bulk packed in bags
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Pimkhuan Hannanta-Anan et al.
ACS synthetic biology, 7(6), 1488-1495 (2018-05-25)
As fast terminators of G-protein coupled receptor (GPCR) signaling, regulators of G-protein signaling (RGS) serve critical roles in fine-tuning second messenger levels and, consequently, cellular responses to external stimuli. Here, we report the creation of an optogenetic RGS2 (opto-RGS2) that
Mathias Foo et al.
ACS synthetic biology, 7(6), 1553-1564 (2018-05-11)
Crop disease leads to significant waste worldwide, both pre- and postharvest, with subsequent economic and sustainability consequences. Disease outcome is determined both by the plants' response to the pathogen and by the ability of the pathogen to suppress defense responses