Key Documents
Z742089
Western Blot Box
size medium, clear, size 11.1 cm × 5.2 cm × 4.7 cm, pk of 5
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
size
medium
packaging
pk of 5
manufacturer/tradename
MIDSCI SG-WBB11
size
11.1 cm × 5.2 cm × 4.7 cm
color
clear
General description
  • Perfect box for every blot
  • Reduce antibody usage
  • Hinged Lid minimizes evaporation
  • For halves, wholes, strips, or minis
No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).
At a price that makes them truly disposable, all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.
Choose from one of the most recent versions:
Certificates of Analysis (COA)