Key Documents
Z742088
MultiBox 6, 6 compartment blot box
width × L × H 4.25 in. × 8.25 in. × 1.375 in., pk of 4
About This Item
UNSPSC Code:
41105300
packaging
pk of 4
width × L × H
4.25 in. × 8.25 in. × 1.375 in.
General description
The MultiBox 6 Western blot Box is a multi-compartment box for Western blot strips. Each of the 6 sections measures 32 X 100mm. The uniformly flat bottom surface allows complete strip coverage with minimal reagent consumption. Made of advanced polystyrene blend, these blot boxes are durable and nearly unbreakable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)