Key Documents
Z741678
Sartorius mLINE® mechanical pipettor Multipacks
Volume: Multiple (10 μl, 200 μl, 300 μl,1,000 μl)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
description
Includes stand and 10 μl, 200 μl, 350 μl and 1000 μl tip trays
feature
8-channel
manual
packaging
pack of 4 pieces
manufacturer/tradename
Sartorius LH-725665
volume
1,000 μL
10 μL
200 μL
300 μL
General description
The mLINE® mechanical pipette Multipacks offer an affordable option to get a set of quality pipettes with useful accessories in a convenient package of your choice. Whether you are renewing your existing pipettes or setting up a new pipetting station, these packs will fulfill your need. Sartorius′ most advanced mechanical pipette family - the mLINE[ - offers excellent ergonomics, performance and safety in manual pipetting. It is particularly designed for repetitive and long-lasting pipetting in order to prevent work related hand, arm and shoulder disorders or Repetitive Strain Injury (RSI). mLINE is recommended by health and safety officers around the globe.
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)