Key Documents
Z741677
Sartorius mLINE® mechanical pipettor Multipacks
Volume: Multiple (20 μl, 100 μl, 200 μl, 1,000 μl, 5,000 μl)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
description
Includes stand and 200 μl x3, 1000 μl and 5000 μl Tip Trays
feature
manual
single-channel
packaging
pack of 5 pieces
manufacturer/tradename
Sartorius LH-725664
volume
1,000 μL
100 μL
20 μL
200 μL
5,000 μL
General description
The mLINE® mechanical pipette Multipacks offer an affordable option to get a set of quality pipettes with useful accessories in a convenient package of your choice. Whether you are renewing your existing pipettes or setting up a new pipetting station, these packs will fulfill your need. Sartorius′ most advanced mechanical pipette family - the mLINE[ - offers excellent ergonomics, performance and safety in manual pipetting. It is particularly designed for repetitive and long-lasting pipetting in order to prevent work related hand, arm and shoulder disorders or Repetitive Strain Injury (RSI). mLINE is recommended by health and safety officers around the globe.
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)