Key Documents
Z741657
Sartorius extended length pipette tips
refill, volume range 0.1-10 μL, sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
sterility
sterile
feature
DNase free
RNase free
barrier: no
endotoxin free
packaging
pack of 960 ea (Racked)
manufacturer/tradename
Sartorius 783211
L
46 mm
maximum volume
10 μL
volume range
0.1-10 μL
General description
Perfect tip fitting and sealing to the pipetter results in the best possible accuracy and precision. Compatible with Optiload feature in Biohit Picus, eLINE®, mLINE® and Proline® Plus pipetters, enabling ergonomic and light tip attachment and ejection.
Other Notes
Browse all pipette tips
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)