Key Documents
Z741637
Sartorius Proline® Plus fixed volume mechanical pippettor
Single Channel, volume 5000 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
manual
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728580
volume
5000 μL
General description
Proline® Plus mechanical pipette family is designed to offer comfort and quality for your every day manual pipetting. While it shares many of the excellent features of mLINE®, it has a personal design and a robust feel for heavier use. Fixed volume pipettes and ideal for when volumes need to be ready-set to avoid errors. ProlineProline® Plus pipettes are an excellent choice for both experienced laboratory professionals and students.
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)