Z741323
Techne® digital Dri-Block® heaters
4 block, max. temp. 100 °C, AC/DC input 240 V AC, Schuko and UK plug
About This Item
Recommended Products
AC/DC input
240 V, Schuko plug (AC)
material
aluminum plate
feature
CE compliant
manufacturer/tradename
Techne DB100 / 4
parameter
100 °C max. temp.
capacity
, holds 4 blocks
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Small, light, compact footprint - flat top surface allows units to be stacked in storage.
- Can hold any combination of aluminium insert blocks, depending on the model chosen.
- Digital models enable highly accurate temperature precision.
- Temperature range from ambient + 5°C to 200°C depending on the model chosen.
- Fast heat-up rate: 25°C to 100°C in 7-9 minutes, depending on the model chosen.
- Advanced temperature control. Temperature stability at 37°C ± 0.1°C.
- Outer casing is cool to the touch, even at maximum operating temperature.
- Bright orange 5-digit LED digital display for fast and accurate setting of temperature and timer.
- Temperature range from ambient to 100°C or 200°C, depending on the model chosen.
- Accurate, reproducible, rapid and safe heating of your samples.
- High quality, precision-engineered blocks provide excellent thermal contact.
- Choice of models with 2, 3 or 4 interchangeable blocks.
- Versatile range of interchangeable heating blocks to fit all popular sample tubes and plates.
- Full range of models and options for basic and advanced applications.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service