Key Documents
Z741111
Eppendorf® ep Dualfilter T.I.P.S.® SealMax
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
material
yellow
sterility
sterile (PCR clean/sterile/pyrogen-free)
feature
DNase free
RNase free
Low binding, filter, RNA or DNA free, pcr, low retention etc.
packaging
pack of 960 ea
rack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030 077.822 EU
Eppendorf® 030077822 US
maximum volume
100 μL
binding type
low binding surface
General description
The new violet and white filter for the ep Dualfilter T.I.P.S.® SealMax filter tips combine a reliable contamination protection for your pipette from your sample liquid with virtually 100% protection from aerosols1 and biomolecules.
This new Dualfilter closes immediately when it comes into contact with the overpipetted sample without inhibiting valuable material. Additionally drops and splashes are retained by the white layer, the violet layer forms the highly-effective barrier against sample liquid.
Further information and certification on specific sample recovery features, PCR inhibition and aerosol protection are available on your local Eppendorf website.
Product Applications:
  • DNA applications (e.g. PCR)
  • RNA applications (e.g. Gene expression analysis)
  • Protein applications (e.g. Antibody Research)
  • Cell Culture applications (e.g. Media)
  • Applications with radio actives
  • All applications which use aerosol1-binding liquids
Product Features:
  • Reliable protection from liquids
  • Reliable protection from aerosols1 and biomolecules
  • No PCR inhibiting additives
  • Protection of your pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean / Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)