Key Documents
Z740463
Isohelix Dri-Capsules
with 50 SK1 swab kits, pkg of 50 ea
About This Item
UNSPSC Code:
41100000
packaging
pkg of 50 ea
General description
Isohelix Dri-Capsules
Designed for use with Isohelix SK-1 swabs, these silica gel capsules offer an easy to use alternative to chemical stabilisation for long term stability of the buccal DNA on the swab head prior to isolation.
Features:
  • Long term stability at room temperature
  • Liquid free alternative to DSK Kit
  • Easy to handle by patients and professionals
  • Designed for use with SK-1 swab packs
  • Ideal for remote sample collection
Long term stability studies show that the stability of DNA samples on SK1 swabs stored with Isohelix Dri-Capsules is maintained over a period of at least 3 years. The ongoing study is expected to confirm an increased period of stabilization well in excess of the 3 years tested so far.
Choose from one of the most recent versions:
Certificates of Analysis (COA)