Z740458
Isohelix DNA Buccal Swabs, ethylene oxide treated
3 swabs per wrapper, pkg of 600 ea
About This Item
Recommended Products
packaging
pkg of 600 ea
General description
Isohelix DNA swabs are suitable for both human and veterinary use and offer significant advantages over other DNA swab designs in terms of efficiency of cell collection by the unique swab matrix, which combined with a quick release surface, maximizes yields of DNA. Swab batches are ethylene oxide treated and routinely tested for human DNA contamination.
Features:
- High yielding swabs for buccal cell DNA collection
- Unique swab matrix greatly improves DNA yields
- Alternative to blood collection
- Suitable for human and veterinary use
- Easy to handle and quick to use
- Available in single and twin sterile packs
Applications:
- Research Genotyping Applications
- Veterinary Genotyping and Diagnostics
- Paternity and Heredity Services
- Forensics and DNA Population Studies
Choose from one of the most recent versions:
Certificates of Analysis (COA)