MilliporeSigma
All Photos(4)
Key Documents
Z740360
Eppendorf® Reference® 2 Variable Volume Pipettor
volume - 0.1-1 mL, 0.5-5 mL, 1-10 mL, pack of 3 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
electronic: no
single-channel
packaging
pack of 3 ea
manufacturer/tradename
Eppendorf® 4924000924
volume measuring range
0.1-1 mL
0.5-5 mL
1-10 mL
volume
10 mL
General description
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Packaging
Includes: 3 adjustable-volume pipettes (1-10 mL, 0.5-5 mL, 100-1,000 mL), 1 full box of Eppendorf pipette tips for each pipette volume (excludes 5 mL and 10 mL tips)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)