Key Documents
Z740354
Nunc® flasks, Nunclon® Δ surface
surface area × working volume 80 cm2 × 30 mL, vent/close cap, straight neck, sterile; γ-irradiated
Synonym(s):
cell culture flask, treated flask
About This Item
UNSPSC Code:
41122104
Pricing and availability is not currently available.
material
screw top blue high-density polyethylene cap
screw top cap
straight neck transparent polystyrene flask
sterility
sterile; γ-irradiated
feature
with cap (vent)
packaging
case of 50 ea (10 packs x 5 flasks)
manufacturer/tradename
Nunc 153732
flask working volume
30 mL
surface area × working volume
80 cm2 × 30 mL
working volume
30 mL
binding type
Nunclon Delta surface
General description
Nunc Flasks with Nunclon Delta treated surface are designed for enhanced cell attachment, growth and differentiation. Vent/close caps of sterile high-density polyethylene (HDPE) provide sufficient gas exchange.
Includes:
  • Caps
  • Nunclon Delta Certification including testing on 4 different cell types is provided in each case
  • Short, wide necks allow for easy access
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)