Z740289
RotoFlex® Plus bench top tube rotator
AC/DC input 120 V AC
Synonym(s):
Western Blot hybridization, sample mixing, tube rotator
About This Item
Recommended Products
feature
CSA approved
Unit is supplied w/rotisseries, 1 pack of each size
manufacturer/tradename
Cole Parmer 439740
General description
The easy to operate key pad allows for speed adjustments between 10 - 40 rpm. In addition to the variable speed feature the unit can also be operated in an oscillating mode. In oscillation mode the rotisseries rotate 40°, pause, then oscillate back and forth five times before rotating another 40° to repeat the process. After oscillating for 360°, the rotisserie rotates a complete turn before reverting back to the oscillation cycle.
The RotoFlex Plus can also be used with a magnetic platform (sold separately) to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane & antibody solution to the plate using the metal strips provided.
Features:
To change the rotation direction - simply touch the rotisserie paddle while in motion and the RotoFlex Plus automatically begins to rotate in the opposite direction. The RotoFlex Plus can be used in incubators & cold room settings. Unit is supplied with base unit and rotisseries (1 pack of each size).
Ideal for:
- Hybridization
- DNA Purification
- Sample mixing/resuspension
- Protein Binding
- Immunoprecipitation
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)