Key Documents
Z740124
BRAND® pipette tips, racked, TipStack
volume 0.5-20 μL, BIO-CERT
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
colorless polypropylene tip
polypropylene
sterility
sterile; autoclaved
feature
RNase and DNase free
endotoxin free
BIO-CERT
packaging
pack of 960 ea (10 racks of 96 + 2 boxes)
manufacturer/tradename
BRAND 732264
maximum volume
20 μL
volume
0.5-20 μL
General description
TipStack, non-sterile and sterile, BIO-CERT®
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
A tip tower containing 5 filled tip-trays and a TipBox constitute the new, space-saving refill system for 20 μL, 200 μL and 1000 μL tips. Tightly sealing spacers prevent the tips from getting stuck together, and ensure them to be free from DNA, RNases, endotoxins and ATP.
The sterile TipStacks (BIO-CERT® quality) are supplied with a transfer aid for contamination-free use in a previously sterilized TipBox. Each packaging unit contains 2 TipStacks.
  • All components are recyclable
  • Reduced amount of waste
  • Sterilizable and reusable TipBox
  • High purity of the pipette tips and filter tips
  • Space-saving design
Other Notes
Browse all pipette tips
Legal Information
BACH-EI is a trademark of Sigma-Aldrich Co. LLC
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)