Key Documents
Z740079
BRAND® pipette tips low retention, racked, TipStack
volume 0.5-20 μL, BIO-CERT®
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
colorless polypropylene tip
polypropylene
sterility
sterile; autoclaved
feature
RNase and DNase free
endotoxin free
BIO-CERT®
packaging
pack of 960 ea (10 racks of 96 + 2 boxes)
manufacturer/tradename
BRAND 732364
maximum volume
20 μL
volume
0.5-20 μL
binding type
low binding surface
General description
A tip tower containing 5 filled tip-trays and a TipBox constitute the new, space-saving refill system for 20 μL, 200 μL and 1000 μL tips. Tightly sealing spacers prevent the tips from getting stuck together, and ensure them to be free from DNA, RNases, endotoxins and ATP.
The sterile TipStacks (BIO-CERT® quality) are supplied with a transfer aid for contamination-free use in a previously sterilized TipBox. Each packaging unit contains 2 TipStacks.
  • All components are recyclable
  • Reduced amount of waste
  • Sterilizable and reusable TipBox
  • High purity of the pipette tips and filter tips
  • Space-saving design
Ultra Low Retention-Pipette tips and Filter tips (ULR)The surfaces of the Ultra Low Retention tips are produced through a special, patented physicochemical process. The homogeneous, defect-free surface thus produced has extremely low surface tension - over 50% less than PTFE (see table). This significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.
  • Ideal for biological samples that contain detergents such as Triton X-100, SDS, Tween etc
  • No additives that can be leached out! No siliconization of the surface!
  • High chemical resistance. Ideal for working with solvents
  • The tips can be autoclaved at 121°C (2 bar) without damaging the material properties
Other Notes
Browse all pipette tips
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
Choose from one of the most recent versions:
Certificates of Analysis (COA)