A Longer Working Distance for when you can't get close enough to focus on what you want to see. They are included with Jenco brand inverted microscopes.
Z738166
Jenco™ stereo microscopes, dissecting
AC/DC input 220 V AC, binocular 20x-40x, illumination: halogen top; fluorescent bottom
Synonym(s):
dissecting microscopes, microscopes, stereo microscopes
About This Item
Recommended Products
description
3 Plug
EU 2-prong plug
manufacturer/tradename
Jenco ST-F804-2
General description
Their illuminated stands let you choose light from three directions - above, below or both, to help observe opaque or transparent samples. Solid, all metal focusing mechanisms will endure student use well beyond the five year warranty.
Teachers like them because the eyepieces are locked and don′t walk, the all metal frames are built to last and the low voltage bulbs last for hundreds of hours and are easy to replace. Even models with rechargeable illuminators are available.
- Affordable
- Portable
- Durable
- Easy to use
- Five Year warranty
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
What is a LWD objective?
1 answer-
Helpful?
-
-
Do microscopes just magnify things?
1 answer-
No, microscopes both magnify and clarify. They allow us to see small objects clearly.
Helpful?
-
-
What is a Micrometer on a Jenco™ microscope?
1 answer-
The same as a reticle, only more specific to a group of lines used as a scale for measuring.
Helpful?
-
-
What is resolution in microscopy?
1 answer-
Bringing fine details into focus.
Helpful?
-
-
What is the difference between achromatic, semi-planachromatic, and planachromatic objectives on a Jenco™ microscope?
1 answer-
Each type of objective is color corrected to a higher degree than the previous one. In everyday terms, achromatic objectives afford approximately 80% of the field of view in focus from the center out to the edges. Semi-planachromatic afford approximately 90% and planachromatic afford approximately 100%. The better the color correction, the more cost.
Helpful?
-
-
Do Jenco™ microscopes come with built in lighting?
1 answer-
All microscopes come with their own attached lighting except the stereozoom boomstand and polestand.
Helpful?
-
-
On the Jenco™ microscope, what is a light blue filter used for?
1 answer-
Blue filters are used to change the color of light from the bulb to a cleaner white light.
Helpful?
-
-
On Jenco dual illuminated stereo microscopes, where is the fluorescent light located?
1 answer-
The light is on the bottom or base of the microscope.
Helpful?
-
-
What is a Nosepiece or turret on a Jenco™ microscope?
1 answer-
The part of the microscope where the objectives are attached. It is either reversed, with the objectives pointing away from the operator giving better access to the sample, or standard with objectives pointing toward the operator.
Helpful?
-
-
What Jenco™ microscopes can be used wiith video or digital camera equipment?
1 answer-
We recommend only those models with trinocular heads because they are more stable and we have adapters to fit the trinocular tube, not the eyepiece, Jenco Model BC-311 (Sigma nos. Z735086 for 115V or Z737062 for 220V).
Helpful?
-