Key Documents
Z738050
Jenco stereo microscopes, dissecting
AC/DC input 220 V AC, binocular 10x-40x, illumination: halogen top; fluorescent bottom
Synonym(s):
dissecting microscopes, microscopes, stereo microscopes
About This Item
UNSPSC Code:
41191506
NACRES:
MA.44
description
3 Plug
EU 2-prong plug
manufacturer/tradename
Jenco ST-F803-2
General description
Dissection and observation of macro specimens is more enlightening with a Jenco Stereo microscope. The large stage allows ample room for sample manipulation but still fits in a storage cabinet.
Their illuminated stands let you choose light from three directions - above, below or both, to help observe opaque or transparent samples. Solid, all metal focusing mechanisms will endure student use well beyond the five year warranty.
Teachers like them because the eyepieces are locked and don′t walk, the all metal frames are built to last and the low voltage bulbs last for hundreds of hours and are easy to replace. Even models with rechargeable illuminators are available.
  • Affordable
  • Portable
  • Durable
  • Easy to use
  • Five Year warranty
Legal Information
Jenco is a trademark of Jenco International, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–10 of 39 Questions  
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  2. What is the difference between monocular, binocular, and trinocular for Jenco™ microscopes?
    1 answer
    1. Monocular microscopes have a single eyepiece, binocular microscopes have two eyepieces, and trinocular microscopes have three eyepieces. The trinocular third eyepiece is generally used for photography and video applications.
      Helpful?
  3. What Jenco™ microscopes can be used wiith video or digital camera equipment?
    1 answer
    1. We recommend only those models with trinocular heads because they are more stable and we have adapters to fit the trinocular tube, not the eyepiece, Jenco Model BC-311 (Sigma nos. Z735086 for 115V or Z737062 for 220V).
      Helpful?
  4. What is an Objective on a Jenco™  microscope?
    1 answer
    1. The part of the microscope that magnifies and resolves the image. It is a lens closest to the specimen. A polished chrome plated component with a barrel shape and threads on one end for threading into the nosepiece.
      Helpful?
  5. On a Jenco™  microscope, what does Kohler Illumination do?
    1 answer
    1. Kohler illumination produces an even field of illumination over the whole specimen.
      Helpful?
  6. What is a Stereo Zoom microscope?
    1 answer
    1. A stereo microscope as described above that changes magnification like a zoom lens on a camera, allowing the specimen to stay in focus as magnification changes within a range.
      Helpful?
  7. What is a Condenser on a Jenco microscope?
    1 answer
    1. The piece closest to the light source designed to collect and condense the light and then to focus it onto the specimen.
      Helpful?
  8. On the Jenco™ microscope, what is a light blue filter used for?
    1 answer
    1. Blue filters are used to change the color of light from the bulb to a cleaner white light.
      Helpful?
  9. Do Jenco™ microscopes have the option of increasing magnification through eyepieces?
    1 answer
    1. Yes. All compound microscopes have a 15X option and the Stereo Zoom microscopes have a 15X and 20X option.
      Helpful?
  10. What is a LWD objective?
    1 answer
    1. A Longer Working Distance for when you can't get close enough to focus on what you want to see. They are included with Jenco brand inverted microscopes.
      Helpful?
1–10 of 39 Questions  