We do not suggest using a 35mm Camera as we have found that a digital camera provides a clearer picture.
Z735841
Jenco™ stereo microscopes, dissecting
AC/DC input 115 V AC, binocular 10x-20x, illumination: halogen top; fluorescent bottom
Synonym(s):
dissecting microscopes, microscopes, stereo microscopes
About This Item
Recommended Products
description
3 Plug
manufacturer/tradename
Jenco ST-F801
availability
not available in EU
General description
Their illuminated stands let you choose light from three directions - above, below or both, to help observe opaque or transparent samples. Solid, all metal focusing mechanisms will endure student use well beyond the five year warranty.
Teachers like them because the eyepieces are locked and don′t walk, the all metal frames are built to last and the low voltage bulbs last for hundreds of hours and are easy to replace. Even models with rechargeable illuminators are available.
- Affordable
- Portable
- Durable
- Easy to use
- Five Year warranty
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Can you use a 35mm camera with your Jenco™ microscopes?
1 answer-
Helpful?
-
-
What is a Parfocal on a Jenco microscope?
1 answer-
Parfocal means the center of your specimen will stay in focus as you change magnification.
Helpful?
-
-
What is a Stage or Mechanical stage on a Jenco™ microscope?
1 answer-
A flat plate for placing the slide/specimen with controls to easily move the sample both vertically and horizontally in small increments.
Helpful?
-
-
What objectives can you use immersion oil with on a Jenco™ microscope?
1 answer-
Only those objectives designed for it with an oil specification clearly marked on it's barrel (100X OIL), otherwise the oil creeps up into the microscope causing problems.
Helpful?
-
-
How do you use immersion oil on a Jenco™ microscopes?
1 answer-
Put a drop of oil on top of the slide's cover slip; move the objective down until it touches the oil and you have the specimen in focus.
Helpful?
-
-
On the Jenco™ microscope, what is a light blue filter used for?
1 answer-
Blue filters are used to change the color of light from the bulb to a cleaner white light.
Helpful?
-
-
Do your Jenco™ microscopes come with rubber eyecups?
1 answer-
Yes.
Helpful?
-
-
What is a Stereo microscope?
1 answer-
A microscope that uses two light paths to give a 3-D view of a large specimen. Mainly used for macro sized or solid samples. Limited magnification range, 10x to 200x.
Helpful?
-
-
What is a Nosepiece or turret on a Jenco™ microscope?
1 answer-
The part of the microscope where the objectives are attached. It is either reversed, with the objectives pointing away from the operator giving better access to the sample, or standard with objectives pointing toward the operator.
Helpful?
-
-
What is a compound microscope?
1 answer-
A microscope with one light path going through compound parts providing high magnification (40x to 1,000x or more). Jenco Model BC-211 (Sigma nos. Z734977 for 115V or Z736945 for 220V) for example.
Helpful?
-