Yes. All compound microscopes have a 15X option and the Stereo Zoom microscopes have a 15X and 20X option.
Z735744
Jenco™ compound microscopes
AC/DC input 115 V AC, brightfield, achromatic lens, binocular, mechanical stage; LED or other illumination
Synonym(s):
compound microscopes, microscopes
About This Item
description
3-plug
manufacturer/tradename
Jenco CP135-B4
availability
not available in EU
General description
- Illumination can be a mirror, a rechargeable LED, an incandescent bulb or halogen bulb
- Eye pieces come in monocular heads, or teaching heads for assisting students in finding specimens and also binocular heads for easy viewing with both eyes
- Objectives are offered in multiple quality and quantity levels
- Video systems are available to project images on a screen
- Industry leading 5 year warranty
Choose from different frames for the right reason and teach science with the right equipment for your students.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Do Jenco™ microscopes have the option of increasing magnification through eyepieces?
1 answer-
Helpful?
-
-
What is a Reticle on a Jenco™ microscope?
1 answer-
A group of lines, shapes, circles, or angles that are inserted in the eyepiece (usually on a disc) that provide either a standard of measurement or a focal point for the observer.
Helpful?
-
-
What objectives can you use immersion oil with on a Jenco™ microscope?
1 answer-
Only those objectives designed for it with an oil specification clearly marked on it's barrel (100X OIL), otherwise the oil creeps up into the microscope causing problems.
Helpful?
-
-
On a Jenco™ microscope, what does Kohler Illumination do?
1 answer-
Kohler illumination produces an even field of illumination over the whole specimen.
Helpful?
-
-
Do Jenco™ microscopes come with built in lighting?
1 answer-
All microscopes come with their own attached lighting except the stereozoom boomstand and polestand.
Helpful?
-
-
What is the difference between a Stereo and a Dissecting microscope?
1 answer-
There is no difference. Those are different names for the same microscope.
Helpful?
-
-
What does the working distance on a Jenco™ microscope refer to?
1 answer-
The working distance is measured from the end of the objective to the top of the specimen when the specimen is in focus.
Helpful?
-
-
What is the difference between achromatic, semi-planachromatic, and planachromatic objectives on a Jenco™ microscope?
1 answer-
Each type of objective is color corrected to a higher degree than the previous one. In everyday terms, achromatic objectives afford approximately 80% of the field of view in focus from the center out to the edges. Semi-planachromatic afford approximately 90% and planachromatic afford approximately 100%. The better the color correction, the more cost.
Helpful?
-
-
What is an Objective on a Jenco™ microscope?
1 answer-
The part of the microscope that magnifies and resolves the image. It is a lens closest to the specimen. A polished chrome plated component with a barrel shape and threads on one end for threading into the nosepiece.
Helpful?
-
-
What is a Stage micrometer on a Jenco™ microscope?
1 answer-
A graduated scale placed on the stage to be used as a standard of measurement for the specimen.
Helpful?
-