Since achromatic objectives are best corrected spherically for green light, a green filter improves the performance of your achromatic objectives.
Z735515
Jenco™ compound microscopes
AC/DC input 115 V AC, brightfield, achromatic lens, monocular, mechanical stage; LED incandescent or fluorescent
Synonym(s):
compound microscopes, microscopes
About This Item
description
3-plug
manufacturer/tradename
Jenco CP135-M3
availability
not available in EU
General description
- Illumination can be a mirror, a rechargeable LED, an incandescent bulb or halogen bulb
- Eye pieces come in monocular heads, or teaching heads for assisting students in finding specimens and also binocular heads for easy viewing with both eyes
- Objectives are offered in multiple quality and quantity levels
- Video systems are available to project images on a screen
- Industry leading 5 year warranty
Choose from different frames for the right reason and teach science with the right equipment for your students.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
On the Jenco™ microscope, what is a green filter used for?
1 answer-
Helpful?
-
-
Why do some objectives on a Jenco™ microscope have retractable lenses?
1 answer-
This feature provides protection against damage caused by a collision of the objective and the slide.
Helpful?
-
-
What objectives can you use immersion oil with on a Jenco™ microscope?
1 answer-
Only those objectives designed for it with an oil specification clearly marked on it's barrel (100X OIL), otherwise the oil creeps up into the microscope causing problems.
Helpful?
-
-
Can you use a 35mm camera with your Jenco™ microscopes?
1 answer-
We do not suggest using a 35mm Camera as we have found that a digital camera provides a clearer picture.
Helpful?
-
-
Do microscopes just magnify things?
1 answer-
No, microscopes both magnify and clarify. They allow us to see small objects clearly.
Helpful?
-
-
What is a Stage micrometer on a Jenco™ microscope?
1 answer-
A graduated scale placed on the stage to be used as a standard of measurement for the specimen.
Helpful?
-
-
What do the numbers on the outside of the barrel of the objective on a Jenco™ microscope stand for?
1 answer-
They identify five specifications: magnification (60X), aberration correction (PlanAchro), coverglass thickness recommended (0.17), numerical aperture (.85), and specialized optical properties (Phase, (PHP), fluorite (FL), etc.
Helpful?
-
-
What is a compound microscope?
1 answer-
A microscope with one light path going through compound parts providing high magnification (40x to 1,000x or more). Jenco Model BC-211 (Sigma nos. Z734977 for 115V or Z736945 for 220V) for example.
Helpful?
-
-
Do Jenco™ microscopes have the option of increasing magnification through eyepieces?
1 answer-
Yes. All compound microscopes have a 15X option and the Stereo Zoom microscopes have a 15X and 20X option.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-