Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z732001

Hettich® fixed angle rotor, 12 place

for MIKRO 220/220R Centrifuge, Fixed Angle Rotor, 12 place (w/ lid)

Synonym(s):

Hettich® centrifuge rotors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

feature

to hold 12 x 1.5/2.2 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 2218-A

parameter

25,718 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models MIKRO 220/220R

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service