Z729671
Hettich® fixed angle rotor, 12 place
for EBA21 Centrifuge, Fixed Angle Rotor, 12 place
Synonym(s):
Hettich® centrifuge rotors
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103900
feature
RCF 25,718
capacity 1.5 ‑ 2.0 mL
manufacturer/tradename
Hettich 1022
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service