Key Documents
Z724831
Eppendorf® Microcentrifuge 5430 accessories
Fixed Angle Rotor, 30 place, w/ plastic lid
About This Item
UNSPSC Code:
41103900
Product Z724831 is not currently sold in your country. Contact Technical Service
feature
RCF 20,817 (without adapter)
capacity 1.5 ‑ 2.0 mL
lid (plastic)
manufacturer/tradename
Eppendorf® 22654004 (USA)
Eppendorf® 5427 712.003 (European)
Eppendorf® F-45 30-11, 22654004
technique(s)
centrifugation: suitable
General description
  • Standard rotor for Model 5430 spins 30 x 1.5/2.0 ml tubes at molecular grade speed up to 20,817 x g
  • Includes a plastic lid for basic applications
  • Rotor is PTFE-coated for high chemical resistance and easy cleaning
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)