Key Documents
Z722782
Hettich® centrifuge rotors
for EBA 21 Centrifuge, Swing-out Rotor, 12 place (without carriers)
About This Item
UNSPSC Code:
41103900
feature
RCF 2,960
capacity 1.6 ‑ 5 mL
manufacturer/tradename
Hettich 1126
technique(s)
centrifugation: suitable
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)