Key Documents
Z722383
IKA® ULTRA-TURRAX® disperser tubes
ST-20 Stirring tube with built-in stirrer, 10/cs
Synonym(s):
tissue grinders, tissue homogenizers
About This Item
UNSPSC Code:
41121800
packaging
case of 10
pack of 10
manufacturer/tradename
IKA 3599500-10
General description
IKA® ULTRA-TURRAX® Disperser Tubes.
These hermetically sealed tubes prevent the spreading of aerosols or vapours created by mechanical stirring, dispersion or grinding infectious, toxic or odourous substances in open vessels. Disposable and single-use tubes, they avoid the possibility of cross-contamination of samples and remove the need for clean up of disperser tools, stirrer tools, grinding chambers or vessels.
Each tube has a bayonet fitting on the bottom, for easy, secure connection to the tube drive.
Note: Complete with built-in stirrer useful for extractions, soil sample suspensions & solubility testing.
Warning
Stirring, dispersing or grinding may generate heat or cause other changes in the sample that might lead to build up of pressure inside the tube. Use appropriate personal protective equipment when operating and opening tubes.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)