Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z721832

KDS Continuous cycle syringe pump, series 270

AC/DC input 220 V AC, Schuko plug

Synonym(s):

dosing pump, infusion pump, pump infusion

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

CE compliant
2 channels

manufacturer/tradename

KD Scientific 789270-220V

technique(s)

HPLC: suitable
cell culture | mammalian: suitable

volume

0.01-60 mL

suitability

suitable for infusion

Looking for similar products? Visit Product Comparison Guide

General description

Continuous Cycle Syringe Pump. The KDS270 can hold up to four syringes and can cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are withdrawing at the same rate. At the end of the set volume the direction is automatically reversed and the next cycle begins. With the use of 3-way valves the pump can empty and refill syringes in a continuous dispense.
  • Holds four syringes, 10 μL to 60 mL each. With larger syringes the full volume may not be useable. [60 mL syr - 40 mL useable, 30 mL syr - full]
  • Backlit LCD display
  • Knob locks/unlocks drive block for effortless, drag-free adjustment
  • Simple menu-driven setup
  • Select syringe size from displayed table
  • Dispense volume
  • Dispense flow rate
  • Continuous dispense volume display
  • Preset volume control and automatic shut-off
  • Settings can be reviewed or changed during operation
  • Stall detection
  • Choice of unit selection
  • Last settings stored in permanent memory
  • Built-in RS232C interface for computer "daisy chaining" up to 100 pumps
  • TTL interface for foot switch, timer, relay control, outputs for run indicator, valve control

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service