Z721808
KDS Reversible nanoliter syringe pump, series 310
AC/DC input 220 V AC, Schuko plug
Synonym(s):
dosing pump, infusion pump, pump infusion
About This Item
Recommended Products
feature
CE compliant
2 channels
manufacturer/tradename
KD Scientific 789311-220V
volume
0.0005-0.25 mL
suitability
suitable for microfluidics/nanofluidics
General description
- Mini size pump
- 1 to 250 μL microliter syringe
- Simple menu-driven setup with bright display and keypad
- Select syringe size from displayed table
- Dispense volume
- Dispense flow rate
- Infusion or withdrawal
- Dispense volume is displayed and has automatic shutoff at completion
- Easy syringe loading with pusher block fast forward/fast reverse feature for rapid adjustments
- Last settings stored in memory
Choose from one of the most recent versions:
Certificates of Analysis (COA)