Key Documents
Z721794
KDS two syringe nanoliter pump, series 101
AC/DC input 220 V AC, Schuko plug
Synonym(s):
dosing pump, infusion pump, pump infusion
About This Item
UNSPSC Code:
41121500
eCl@ss:
32070500
NACRES:
NB.01
feature
CE compliant
10 channels
manufacturer/tradename
KD Scientific 789101-220V
volume
0.01-60 mL
suitability
suitable for low-volume dispensing
suitable for microdialysis
suitable for microfluidics/nanofluidics
General description
Our dual-syringe infusion pump is ideal for microdialysis and similar applications which require virtually pulseless flow at very low flow rates.
  • Hold 2 syringes, 10 μL to 10 mL each or, a single syringe up to 60 mL
  • Simple menu prompt setup
  • Select syringe size from displayed table
  • Set dispense volume
  • Set dispense flow rate
  • Easy-to-read bright, backlight display
  • Dispense volume displayed and has automatic shutoff at completion
  • Volume and flow rate units (μL/min or mL/min) are selected automatically based on syringe size
  • Last settings stored in permanent memory
  • Button release of drive nut for easy syringe installation
  • Choice of run/stop after a power interruption
Choose from one of the most recent versions:
Certificates of Analysis (COA)