Key Documents
Z721719
KDS Two syringe push-pull pump, series 120
AC/DC input 110 V AC
Synonym(s):
dosing pump, infusion pump, pump infusion
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
2 channels
manufacturer/tradename
KD Scientific 780120-110V
availability
not available in EU
volume
0.01-10 mL
suitability
suitable for cell injection
suitable for perfusion applications
General description
This pump provides simultaneous infusion and withdrawal at the same rate with opposing syringes on the same drive screw.
  • Holds two syringes 10 μL to 10 mL each
  • Back lit LCD display
  • Knob locks/unlocks drive block for effortless drag free adjustment
  • Preset volume control and automatic shut off and stall detection
  • Settings can be reviewed or changed during operation
  • Choice of unit selection
  • Last setting stored in permanent memory
Choose from one of the most recent versions:
Certificates of Analysis (COA)