Z721700
KDS Four syringe push-pull pump series 260
AC/DC input 110 V AC
Synonym(s):
dosing pump, infusion pump, pump infusion
About This Item
Recommended Products
feature
CE compliant
4 channels
manufacturer/tradename
KD Scientific 780260-110V
availability
not available in EU
volume
0.01-60 mL
suitability
suitable for cell injection
Looking for similar products? Visit Product Comparison Guide
General description
Note: When not used in push/pull mode, the pump has all the features of KDS210.
- Holds up to four syringes, 10 μL to 60 mL each. With large syringes, the full volume may not not be usable
- Backlit LCD display
- Knob locks/unlocks drive block for effortless, drag-free adjustment
- Simple menu-driven setup: select syringe size from displayed table dispense volume dispense flow rate
- Continuous dispense volume display
- Preset volume control and automatic shut-off
- Settings can be reviewed or changed during operation
- Stall detection
- Choice of unit selection
- Last settings stored in permanent memory
- Built-in RS-232C interface for computer "daisy chaining" up to 100 pumps
- TTL interface for foot switch, timer, relay control, outputs for run indicator, valve control
- Independent rate and volume settings for infusion and withdrawal
- Multiple mode selection: Withdrawal for push/pull
- Optional multi-step keypad programmable feature
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service