Z721697
KDS two syringe nanoliter pump, series 101
AC/DC input 110 V AC
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
2 channel
manufacturer/tradename
KD Scientific 780101-110V
availability
not available in EU
volume
0.01-10 mL
suitability
suitable for low-volume dispensing
suitable for microdialysis
suitable for microfluidics/nanofluidics
General description
Our dual-syringe infusion pump is ideal for microdialysis and similar applications which require virtually pulseless flow at very low flow rates.
- Hold 2 syringes, 10 μL to 10 mL each or, a single syringe up to 60 mL
- Simple menu prompt setup
- Select syringe size from displayed table
- Set dispense volume
- Set dispense flow rate
- Easy-to-read bright, backlight display
- Dispense volume displayed and has automatic shutoff at completion
- Volume and flow rate units (μL/min or mL/min) are selected automatically based on syringe size
- Last settings stored in permanent memory
- Button release of drive nut for easy syringe installation
- Choice of run/stop after a power interruption
Choose from one of the most recent versions:
Certificates of Analysis (COA)