Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z721263

MicroCaster microarray system

MicroCaster pad (pin support pad), 1/pk

Synonym(s):

microarray system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

packaging

pack of 1

manufacturer/tradename

Whatman 10 485 370

Looking for similar products? Visit Product Comparison Guide

General description

Handheld Microarraying System
The MicroCaster is an economical, entry-level manual microarraying system for principle and pilot studies. With the MicroCaster 8-pin hand tool, samples can be loaded from 96 well or 384 well microtiter plates.

The MicroCaster slide holder accommodates two slides. It has a built-in indexing system that enables precise printing of up to 768 spots in an array of 32 x 24 spots. It is very easy to set up and use with processing time of 5 to 20 minutes per slide.

The MicroCaster is designed for 1-pad FAST Slides with 20 x 51 mm pad size and is compatible with other slide surfaces.

MicroCaster accessories can be used to increase the flexibility of the manual arrayer system by providing accurate source-plate indexing and reliable pin-tool cleaning.

The MicroCaster accessories are compatible with standard 96 well microplates and they reduce hassle with pin-tool cleaning.

Specifications:
  • Number of spots: up to 768
  • Horizontal pitch: x-axis 1250 μm
  • Vertical pitch: y-axis 750 μm
  • Spot size: 500 to 1000 μm
  • Print volume: (varies with buffer and viscosity) 20 to 70 nL

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service