Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z721174

FAST PAK protein array kits

FAST PAK 2 pad protein array kit, 1/pk

Synonym(s):

antibody arrays, protein microarrays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

packaging

kit of 1

manufacturer/tradename

Maine Mfg 10485319

suitability

suitable for (ELISA format (sandwich assay) antibody arrays, protein arrays, carbohydrate arrays, reverse phase (micro-Western) arrays using cell or tissue lysates)

Looking for similar products? Visit Product Comparison Guide

General description

FAST PAK protein array kits provide the necessary components for researchers to conveniently build and process protein microarrays.
FAST PAK kits are available for all FAST Slide formats (1, 2, 8, and 16 pads). Each kit contains 10 FAST Slides, 10 Incubation Chambers, 40 Chamber Covers, 10 ml 2 x Protein Arraying Buffer, 15 ml Protein Array Blocking Buffer, and 125 ml 10 x Protein Array Wash Buffer.
Note: A reusable slider holder is also needed, either FAST Frame for 1 to 4 slides or Chip Clip for 1 slide.
Features and benefits
  • 1, 2, 8, or 16 array pads on each slide
  • Each pad can be processed separately to increase the number of arrays on each slide and reduce sample volume
  • Suitable for multiplex experiments, side-by-side comparisons, and control experiments all on the same slide
  • Reproducible results from slide-to-slide and pad-to-pad
  • Increased protein stability and enhanced signal intensity with FAST PAK array buffer
  • Proprietary protein array blocking buffer ensures optimum signal-to-noise ratios and promotes specific binding
  • Flexibility for detection by fluorescent, chemiluminescent, colorimetric, and radiographic methods

Applications
  • ELISA format (sandwich assay) experiments using antibody arrays
  • Reverse phase (micro-Western) arrays using cell or tissue lysates
  • Purified protein arrays
  • Carbohydrate arrays
  • Lipids and other materials which can be arrayed on nitrocellulose

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service