Z720496
Nunc® EZ Flip™ conical centrifuge tubes
50 mL conical tube, bulk packed
Synonym(s):
centrifuge tubes
About This Item
Recommended Products
material
cap (leakproof cap)
conical bottom tube
polypropylene
sterility
sterile
feature
RCF 9,500 × g
packaging
pack of 25
case of 500
manufacturer/tradename
Nunc 362696
capacity
50 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
General description
Ergonomic design has a hinged-cap system for one-handed opening and closing. Integrated leakproof cap prevents cross contamination and lost closures. Clarified polypropylene combines chemical resistance and high speed tolerance. 15 mL and 50 mL versions are available White writing area and flat caps for sample identification. Printed graduations. Standard dimensions compatible with centrifuge rotors.
- Hinged-cap system with ergonomic design for one-handed opening and closing
- Integrated leakproof cap prevents cross contamination and lost closures
- Clarified polypropylene combines chemical resistance and high speed tolerance
- White writing area and flat caps for sample identification
- Printed graduations
- Standard dimensions compatible with centrifuge rotors
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service