Key Documents
Z720070
Millipore
Millipore® Ziptips
C18, pack of 96
Synonym(s):
pipet tips, pipette tips
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
colorless
colorless tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 96 ea
pack of 96
manufacturer/tradename
Millipore ZTC18S096
maximum volume
10 μL
volume range
0.1-10 μL
General description
Millipore ZipTips: Sample Preparation of Peptides or Proteins Prior to Electrospray/Nanoelectrospray MS
ZipTip Advantages:
  • Single-step desalting, concentration, and purification
  • Fractionate complex samples for more meaningful data
  • Ideal for peptides, proteins, nucleic acids, and more
  • No dead volume for maximum recovery
  • Eliminates time-consuming chromatography
Examples of analyses enhanced by ZipTip sample preparation:
  • Mass spectrometry
  • HPLC (high performance liquid chromatography)
  • Capillary electrophoresis
  • Atomic absorption spectroscopy
The ZipTip is a 10 μL (P-10) pipette tip with a bed of chromatography media fixed at its end such that there is no dead volume. It is intended for purifying and concentrating femtomoles to picomoles of protein, peptide or oligonucleotide samples prior to analysis, providing better data quality. The sample is aspirated and dispensed through ZipTip to bind, wash, and elute. Recovered samples are contaminant-free and eluted in 0.5-4 μL for direct transfer to a mass spectrometer target or vial.
ZipTip C18 is a 10 μL pipette tip with approximately a 0.6 μL bed of chromatography media fixed at its end such that there is no dead volume. ZipTip Micro-C18 is a 10 μL pipette tip with approximately a 0.2 μL bed of chromatography media fixed at its end such that there is no dead volume. Both are ideal for concentrating and purifying peptides, proteins or oligonucleotides in seconds prior to mass spectrometry, HPLC, capillary electrophoresis and other analytical techniques.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)