Z709727
Sartorius eLINE® multichannel electronic pipettor
volume 5-120 μL, 8 channel, AC/DC input 115 V AC (USA plug)
Synonym(s):
Biohit pipettes, electronic pipettes, pipettor
All Photos(2)
About This Item
UNSPSC Code:
41121500
feature
electronic
manufacturer/tradename
Sartorius 730341
availability
not available in EU
volume
5-120 μL
General description
With its easy-to-read display and simple-to-operate keyboard the hard work has been removed for programming Sartorius eLINE of the Biohitfamily. Boasting no fewer than 11 pipetting modes including:
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
- Forward pipetting with optional cycle counting
- Reverse and manual pipetting modes
- Multi-dispensing; Diluting
- Automated Dispensing
- Mixing facility
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
Legal Information
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)