Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z709174

ART® self-sealing barrier pipette tips

ART GEL, volume range 0.5-100 μL, non-sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601

material

colorless tip

sterility

non-sterile

feature

barrier

packaging

pack of 1000 ea (bulk)

manufacturer/tradename

Thermo MBP 2155B

technique(s)

PCR: suitable

volume range

0.5-100 μL

Looking for similar products? Visit Product Comparison Guide

General description

Load Agarose and polyacrylamide gels up to 100 μl

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit Proline-Electronic 10-100 μl
Biohit Proline-Variable 50-200 μl
Eppendorf 4810 10-100 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Research/Reference 10-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 Digital 5-40 μl
Mettler-Toledo VoluMate 10-100 μl
Pipetman P-100 10-100 μl
Rainin EDP 2 10-100ul/20-250 μl
Rainin EDP Plus 10-100ul/20-250 μl
Rainin Pipet Plus 10-100ul/20-200 μl
Rainin Pipetman 10-100ul/20-200 μl
Transferpette (Limited Draw) 20-200 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service