Key Documents
Z709085
ART® non-filtered tip
300U, non-sterile
Synonym(s):
pipet tips, pipette tips
About This Item
UNSPSC Code:
41121600
sterility
non-sterile
feature
DNase free
RNase free
packaging
pack of 960 ea
manufacturer/tradename
Molecular BioProducts 3772-11
dispensing volume measuring range
1-300 μL
maximum volume
300 μL
General description
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
SoftFit-L tip design and sturdy rack make this an ideal choice for multi-channel pipettors.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)