Key Documents
Z708380
Rack, 3-way
natural
About This Item
UNSPSC Code:
41103900
manufacturer/tradename
Heathrow Scientific
color
natural
General description
Polypropylene racks connect to one another on either end. Convenient finger slots are molded onto the rack making them easy to pick up and move. Each of the three tiers accommodates a different sized tube. Lowest level has 24 wells to hold individual 0.2 mL tubes or strips of 8 or 12 tubes. The middle tier has 14 wells to hold 0.5 mL tubes and as many as (12) 1.5mL tubes fit nicely in the top tier. Autoclavable. Available in natural or assorted colors of blue, green, purple, yellow and orange.
Choose from one of the most recent versions:
Certificates of Analysis (COA)